missing translation for 'onlineSavingsMsg'
Learn More

CaCCinh-A01, MedChemExpress

Product Code. 16422626
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16422626

Brand: Medchem Express HY100611

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress CaCCinh-A01 is an inhibitor of both TMEM16A and calcium-activated chloride channel (CaCC) with IC50s of 2.1 and 10 μM, respectively.

CaCCinh-A01 is an inhibitor of both TMEM16A and calcium-activated chloride channel (CaCC) with IC50s of 2.1 and 10 μM, respectively.

Specifications

Chemical Name or Material CaCCinh-A01
Physical Form Solid
CAS 407587-33-1
Color White
Quantity 25 mg
Purity Grade Notes Research
Molecular Formula C18H21NO4S
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : ≥ 50 mg/mL (143.91 mM)
SMILES O=C(C1=C(NC(C2=CC=CO2)=O)SC3=C1CCC(C(C)(C)C)C3)O
Molecular Weight (g/mol) 347.43
Formula Weight 347.43
Percent Purity 99.79%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.