missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CABYR Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA559256

Product Code. 15707121

  • £445.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: GKVSSIHSDQ SDVLMVDVAT SMPVVIKEVP SSEAAEDVMV AAPLVCSGKV LEVQVVNQTS VHVDLGSQPK ENEAEPSTAS SVPLQDEQEP PAY Highest antigen sequence identity to the following orthologs: Mouse - 55%, Rat - 56%.

Spermatozoa gain fertilization capacitation and hyperactivation after residing in the uterus or oviduct. Calcium, cAMP and protein tyrosine phosphorylation are necessary for the molecular mechanisms that allow for this transformation. CABYR (Calcium-binding tyrosine phosphorylation-regulated protein), also known as Fibrousheathin-2 or Cancer/testis antigen 88, is a 493 amino acid protein that is expressed in sperm flagella and exhibits increased tyrosine phosphorylation during capacitation. There are six named isoforms of CABYR that are produced as a result of alternative slicing events. Specifically, isoform 1 is expressed in the testis, while isoform 3 and isoform 5 are expressed in pancreas, brain and various brain tumors. Isoforms 1, 2 and 6 most likely bind calcium, whereas isoforms 3, 4 and 5 probably do not bind calcium.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CABYR
Polyclonal
Unconjugated
CABYR
1700016C01Rik; 4933421A18Rik; Cabyr; CABYRa; CABYRc; CABYRc/d; CABYRe; calcium binding tyrosine phosphorylation regulated; calcium binding tyrosine-(Y)-phosphorylation regulated; calcium binding tyrosine-(Y)-phosphorylation regulated (fibrousheathin 2); calcium-binding protein 86; Calcium-binding tyrosine phosphorylation-regulated protein; calcium-binding tyrosine-(Y)-phosphorylation regulated (fibrousheathin 2); calcium-binding tyrosine-phosphorylation regulated protein; cancer/testis antigen 88; CBP86; CT88; fibrousheathin 2; fibrousheathin II; Fibrousheathin-2; FSP2; FSP-2; testis tissue sperm-binding protein Li 84P; testis-specific calcium-binding protein CBP86
Rabbit
Antigen Affinity Chromatography
RUO
26256
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin)
0.1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
O75952
CABYR
Recombinant protein corresponding to Human CABYR. Recombinant protein control fragment (Product #RP-98250).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.