missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CABS1 Polyclonal antibody specifically detects CABS1 in Human, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | CABS1 |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | DyLight 594 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | C4orf35, calcium-binding protein, spermatid-specific 1, calcium-binding protein, sperm-specific 1, casein-like phosphoprotein, chromosome 4 open reading frame 35, CLPH, FLJ32897, NYD-SP26, Testis development protein NYD-SP26 |
| Host Species | Rabbit |
| Immunogen | Antibody was raised against a 15 amino acid synthetic peptide from near the carboxy terminus of human CLPH. The immunogen is located within the last 50 amino acids of CLPH. Amino Acid Squence: YKEDTSTTETDIFELLKEEPD |
| Purification Method | Peptide affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?