missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CABS1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-77091
This item is not returnable.
View return policy
Description
CABS1 Polyclonal specifically detects CABS1 in Human, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| CABS1 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| C4orf35, calcium-binding protein, spermatid-specific 1, calcium-binding protein, sperm-specific 1, casein-like phosphoprotein, chromosome 4 open reading frame 35, CLPH, FLJ32897, NYD-SP26, Testis development protein NYD-SP26 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 85438 | |
| Human, Rat | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| Western Blot 1-2 ug/ml, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 5 ug/ml | |
| NP_149113 | |
| CABS1 | |
| Antibody was raised against a 15 amino acid synthetic peptide from near the carboxy terminus of human CLPH. The immunogen is located within the last 50 amino acids of CLPH. Amino Acid Squence: YKEDTSTTETDIFELLKEEPD | |
| 0.1 mg | |
| Primary | |
| This antibody will not cross-react with other CLPH family members. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction