missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CaBP8 Polyclonal Antibody

Description
CaBP8 Polyclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | CaBP8 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | CALN1 |
| Gene Accession No. | Q06BI3, Q9BXU9, Q9JJG7 |
| Gene Alias | 9630012C17Rik; CABP8; calcium-binding protein 8; calcium-binding protein CABP8; CALN1; calneuron 1; calneuron I; Calneuron-1; calneuron-I; MNCb-0849 |
| Gene Symbols | CALN1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?