missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CaBP8 Monoclonal Antibody (2G2)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA522701
This item is not returnable.
View return policy
Description
Peptide Sequence: EELDEIREAF RVLDRDGNGF ISKQELGMAM RSLGYMPSEV ELAIIMQRLD MDGDGQVDFD EFMTILGPKL V.
Calcium binding proteins (CaBP) play a crucial role in the calcium-mediated cellular signal transduction pathway in the central nervous system. The CaBP family shares much similarity with CaM I (calmodulin), and it has been shown that CaBP proteins can substitute functionally for, and possibly augment the function of, CaM I. CaBP8, contains two EF-hand domains for calcium binding and shares 70% homology with CaBP7 and 50% homology with CaM I. It negatively regulates Golgi-to-plasma membrane trafficking by interacting with PI4KB and inhibiting its activity. Both CaBP8 and CaBP7 possess a targeting mechanism that is unique amongst the CaBPs that may contribute to differential functional Ca2+-sensing by these family members.
Specifications
| CaBP8 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Q9BXU9 | |
| CALN1 | |
| CALN1 (AAH20200, 36 a.a. ~ 106 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| ELISA, Western Blot | |
| 2G2 | |
| Unconjugated | |
| CALN1 | |
| 9630012C17Rik; CABP8; calcium-binding protein 8; calcium-binding protein CABP8; CALN1; calneuron 1; calneuron I; Calneuron-1; calneuron-I; MNCb-0849 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 83698 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction