missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CABP5 Antibody (016), Alexa Fluor™ 594, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90251AF594
This item is not returnable.
View return policy
Description
CABP5 Monoclonal specifically detects CABP5 in Human samples. It is validated for ELISA.
Specifications
| CABP5 | |
| Monoclonal | |
| Alexa Fluor 594 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 56344 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| 16 | |
| ELISA | |
| CABP3, calcium binding protein 5, calcium-binding protein 3, calcium-binding protein 5 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human CABP5 (Q9NP86; Met1-Arg173). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction