missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CABP2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5103679
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total CABP2.
This gene belongs to a subfamily of calcium binding proteins that share similarity to calmodulin. Like calmodulin, these family members can likely stimulate calmodulin-dependent kinase II and the protein phosphatase calcineurin. Calcium binding proteins are an important component of calcium mediated cellular signal transduction. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq, Jan 2016].
Specifications
| CABP2 | |
| Polyclonal | |
| Unconjugated | |
| CABP2 | |
| CaBP2; calcium binding protein 2; calcium-binding protein 2; DFNB93; RGD1563670 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 29866, 51475 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9JLK4, Q9NPB3 | |
| CABP2 | |
| A synthesized peptide derived from human CABP2(Accession Q9NPB3), corresponding to amino acid residues I66-E116. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction