missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cabozantinib (S)-Malate, Mikromol™
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Description
Cabozantinib (S)-Malate
Specifications
Specifications
| Chemical Name or Material | Cabozantinib (S)-Malate |
| CAS | 1140909-48-3 |
| Analyte or Component Names | Cabozantinib (S)-Malate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C28 H24 F N3 O5 . C4 H6 O5 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Butanedioic acid, 2-hydroxy-, (2S)-, compd. with N-[4-[(6,7-dimethoxy-4-quinolinyl)oxy]phenyl]-N'-(4-fluorophenyl)-1,1-cyclopropanedicarboxamide (1:1), Cabozantinib S-malate, Cabozantinib s-malate, Cometriq, XL 184, Cabozantinib L-malate salt, Cabozantinib (S)-malate |
| InChI Formula | InChI=1S/C28H24FN3O5.C4H6O5/c1-35-24-15-21-22(16-25(24)36-2)30-14-11-23(21)37-20-9-7-19(8-10-20)32-27(34)28(12-13-28)26(33)31-18-5-3-17(29)4-6-18;5-2(4(8)9)1-3(6)7/h3-11,14-16H,12-13H2,1-2H3,(H,31,33)(H,32,34);2,5H,1H2,(H,6,7)(H,8,9)/t;2-/m.0/s1 |
| SMILES | COc1cc2nccc(Oc3ccc(NC(=O)C4(CC4)C(=O)Nc5ccc(F)cc5)cc3)c2cc1OC.O[C@@H](CC(=O)O)C(=O)O |
| IUPAC Name | 1-N-[4-(6,7-dimethoxyquinolin-4-yl)oxyphenyl]-1-N'-(4-fluorophenyl)cyclopropane-1,1-dicarboxamide;(2S)-2-hydroxybutanedioic acid |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction