missing translation for 'onlineSavingsMsg'
Learn More

Cabozantinib S-malate, MedChemExpress

Product Code. 16481026
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16481026

Brand: Medchem Express HY12044

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Cabozantinib S-malate (XL184 S-malate) is a potent multiple receptor tyrosine kinases inhibitor that inhibits VEGFR2, c-Met, Kit, Axl and Flt3 with IC50s of 0.035, 1.3, 4.6, 7 and 11.3 nM, respectively.

Cabozantinib S-malate (XL184 S-malate) is a potent multiple receptor tyrosine kinases inhibitor that inhibits VEGFR2, c-Met, Kit, Axl and Flt3 with IC50s of 0.035, 1.3, 4.6, 7 and 11.3 nM, respectively.

Specifications

Chemical Name or Material Cabozantinib S-malate
CAS 1140909-48-3
Color White
Purity Grade Notes Research
Molecular Formula C32H30FN3O10
Quantity 200 mg
Synonym XL184 S-malateBMS-907351 S-malate
Solubility Information DMSO : ≥ 23 mg/mL (36.19 mM) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C([C@H](CC(O)=O)O)O.O=C(NC1=CC=C(C=C1)OC2=CC=NC3=CC(OC)=C(C=C23)OC)C4(CC4)C(NC5=CC=C(C=C5)F)=O
Molecular Weight (g/mol) 635.59
Formula Weight 635.59
Percent Purity 99.29%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.