missing translation for 'onlineSavingsMsg'
Learn More

Cabotegravir, TRC

Product Code. 17975035
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17975035 5 mg 5mg
17963345 10 mg 10mg
17952305 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17975035 Supplier Toronto Research Chemicals Supplier No. TRCC0501005MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1051375-10-0
Molecular Formula C19 H17 F2 N3 O5
Molecular Weight (g/mol) 405.35
Synonym Oxazolo[3,2-a]pyrido[1,2-d]pyrazine-8-carboxamide, N-[(2,4-difluorophenyl)methyl]-2,3,5,7,11,11a-hexahydro-6-hydroxy-3-methyl-5,7-dioxo-, (3S,11aR)-, (3S,11aR)-N-[(2,4-Difluorophenyl)methyl]-2,3,5,7,11,11a-hexahydro-6-hydroxy-3-methyl-5,7-dioxooxazolo[3,2-a]pyrido[1,2-d]pyrazine-8-carboxamide, (3s,11Ar)-n-(2,4-difluorobenzyl)-6-hydroxy-3-methyl-5,7-dioxo-2,3,5,7,11,11a-hexahydrooxazolo[3,2-a]pyrido[1,2-d]pyrazine-8-carboxamide, Cabotegravir, GSK 1265744A, GSK 744, GSK 1265744A
IUPAC Name (3R,6S)-N-[(2,4-difluorophenyl)methyl]-10-hydroxy-6-methyl-8,11-dioxo-4-oxa-1,7-diazatricyclo[7.4.0.03,7]trideca-9,12-diene-12-carboxamide
SMILES C[C@H]1CO[C@@H]2CN3C=C(C(=O)NCc4ccc(F)cc4F)C(=O)C(=C3C(=O)N12)O

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C19H17F2N3O5/c1-9-8-29-14-7-23-6-12(16(25)17(26)15(23)19(28)24(9)14)18(27)22-5-10-2-3-11(20)4-13(10)21/h2-4,6,9,14,26H,5,7-8H2,1H3,(H,22,27)/t9-,14+/m0/s1
Formula Weight 405.1136
Percent Purity >95%
Chemical Name or Material Cabotegravir
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.