missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cabotegravir, MedChemExpress
Description
Cabotegravir is primarily metabolized by uridine diphosphate glucuronosyltransferase (UGT) 1A1, with low potential to interact with other antiretroviral drugs (ARVs). Cabotegravir can be used to research AIDS.
Specifications
Specifications
| Chemical Name or Material | Cabotegravir |
| Physical Form | Powder |
| CAS | 1051375-10-0 |
| Color | Light Yellow |
| Quantity | 10 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C19H17F2N3O5 |
| For Use With (Application) | COVID-19-anti-virus |
| Synonym | GSK-1265744 S/GSK1265744 |
| Solubility Information | DMSO : 16.67 mg/mL (41.12 mM; Need ultrasonic) |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?