missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CABLES1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5103206
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total Ik3-1.
Cyclin-dependent kinase binding protein. Enhances cyclin-dependent kinase tyrosine phosphorylation by nonreceptor tyrosine kinases, such as that of CDK5 by activated ABL1, which leads to increased CDK5 activity and is critical for neuronal development, and that of CDK2 by WEE1, which leads to decreased CDK2 activity and growth inhibition. Positively affects neuronal outgrowth. Plays a role as a regulator for p53/p73-induced cell death.
Specifications
| CABLES1 | |
| Polyclonal | |
| Unconjugated | |
| CABLES1 | |
| CABL1; CABLES; CABLES1; cdk3-binding protein ik3-1; CDK5 and Abl enzyme substrate 1; CDK5 and ABL1 enzyme substrate 1; HsT2563; ik3-1; interactor with CDK3 1; interactor-1 with cdk3 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 63955, 91768 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8TDN4, Q9ESJ1 | |
| CABLES1 | |
| A synthesized peptide derived from human CABLES1(Accession Q8TDN4), corresponding to amino acid residues L568-P618. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction