Learn More
Fisherbrand™ Cable
Designed for use with TRITON II trinocular microscopes. Fisherbrand™ Cables feature a UK plug and Euro connector.
£30.70
Specifications
Type Cable
Color Black
Length (Metric) 2 m
Quantity 1 Ea.
View More Specs
Products 1
Product Code Brand Includes Price Quantity & Availability  
Product Code Brand Includes Price Quantity & Availability  
12914371
View Documents Promotion Details
Fisherbrand™
UK Plug and Euro Connector
£30.70
Each
View alternative products
Save Up to 
»
Estimated Shipment: 25-07-2024
to see stock.
Add to basket
Description
Description
Trinocular configuration allows for the addition of a camera for digital imaging or video camera.
The diamond shaped base provides stability and hand support whilst samples are manipulated on the stage. The zoom ratio of 1x to 4x gives a magnification of 10x to 40x with the standard 10x eyepieces; both with dioptre compensation. Additional eyepieces and corrective lenses allow for a total magnification range from 2.5 to 120x. Both light sources have variable intensity control. The unit comes with frosted glass stage, a blue filter and a heat filter for the incident light source to prevent overheating of the sample.
Robust stereo zoom microscope
  • Integral single piece column stand eliminates vibration and image distortion
  • Ergonomic diamond shaped base
  • Total magnification range 2.5x to 120x
  • Variable intensity transmitted and incident light sources
Specifications
Specifications
Cable
Black
2 m
1 Ea.