missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Cabergoline
GREENER_CHOICE

Product Code. 18772694
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18772694

Brand: Tocris Bioscience™ 2664/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective D2-like agonist

Cabergoline is a selective D2-like dopamine receptor agonist (Ki values are 0.7, 1.5, 9.0 and 165 nM for D2, D3, D4 and D5 receptors respectively) that also displays high affinity for several serotonin receptor subtypes (Ki = 1.2 - 20.0 nM for 5-HT1A, 5-HT1D, 5-HT2A and 5-HT2B). Inhibits secretion of prolactin and growth hormone and reverses levodopa -induced dyskinesias in Parkinsonian monkeys.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 81409-90-7
Molecular Formula C26H37N5O2

Specifications

Quantity 10 mg
Formula Weight 451.6
Purity >99%
Molecular Weight (g/mol) 451.615
Chemical Name or Material Cabergoline
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.