missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cabergoline, MedChemExpress
Description
Cabergoline is an ergot derived-dopamine D2-like receptor agonist that has high affinity for D2, D3, and 5-HT2B receptors (Ki=0.7, 1.5, and 1.2, respectively).
Specifications
Specifications
| Chemical Name or Material | Cabergoline |
| CAS | 81409-90-7 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C26H37N5O2 |
| Quantity | 5 mg |
| Synonym | FCE-21336 |
| Solubility Information | DMSO : 250 mg/mL (553.59 mM; Need ultrasonic) |
| SMILES | C=CCN1[C@@]2([H])[C@@](C[C@H](C1)C(N(CCCN(C)C)C(NCC)=O)=O)([H])C3=CC=CC4=C3C(C2)=CN4 |
| Molecular Weight (g/mol) | 451.6 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?