missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Immunogen sequence: EMKRLDQEQG PEESQPETDT SDMDPLVGFT IEDVKSVLDQ VTDDILIGIQ TEINEKLQIQ EEAFNARIE Highest antigen sequence identity to the following orthologs: Mouse - 64%, Rat - 67%.
CABCOCO1 (C10orf107) is a calcium-binding protein that may be involved in the control of sperm flagellar movement.
Specifications
Specifications
| Antigen | CABCOCO1 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | CABCOCO1 |
| Gene Accession No. | Q8IVU9 |
| Gene Alias | bA63A2.1; C10orf107; CABCOCO1; chromosome 10 open reading frame 107; Ciliary-associated calcium-binding coiled-coil protein 1; RGD1306739; similar to RIKEN cDNA 1700040L02; uncharacterized protein C10orf107; uncharacterized protein C10orf107 homolog |
| Gene Symbols | CABCOCO1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?