missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CAB39L Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92451-0.1ml
This item is not returnable.
View return policy
Description
CAB39L Polyclonal antibody specifically detects CAB39L in Human, Mouse samples. It is validated for Western Blot
Specifications
| CAB39L | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| Antigen MLAA-34, bA103J18.3, calcium binding protein 39-like, calcium-binding protein 39-like, FLJ12577, MO25-BETA, Mo25-like protein, MO2L, RP11-103J18.3, sarcoma antigen NY-SAR-79, U937-associated antigen | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-337 of human CAB39L (NP_112187.2). MKKMPLFSKSHKNPAEIVKILKDNLAILEKQDKKTDKASEEVSKSLQAMKEILCGTNEKEPPTEAVAQLAQELYSSGLLVTLIADLQLIDFEGKKDVTQIFNNILRRQIGTRSPTVEYISAHPHILFMLLKGYEAPQIALRCGIMLRECIRHEPLAKIILFSNQFRDFFKYVELSTFDIASDAFATFKDLLTRHKVLVADFLEQNYDTIFEDYEKLLQSENYVTKRQSLKLLGELILDRHNFAIMTKYISKPENLKLMMNLLRDKSPNIQFEAFHVFKVFVASPHKTQPIVEILLKNQPKLIEFLSSFQKERTDDEQFADEKNYLIKQIRDLKKTAP | |
| 0.1 mL | |
| Cancer, Endocrinology | |
| 81617 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction