missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CAB39 Recombinant Rabbit Monoclonal Antibody (24GB6950)
Rabbit Recombinant Monoclonal Antibody
£126.00 - £434.00
Specifications
| Antigen | CAB39 |
|---|---|
| Clone | 24GB6950 |
| Concentration | 0.3 mg/mL |
| Content And Storage | -20°C |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30276627
|
Invitrogen™
MA556995 |
100 μL |
£434.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30275929
|
Invitrogen™
MA556996 |
20 μL |
£126.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CAB39 Recombinant Monoclonal Antibody for Western Blot
Peutz-Jeghers Syndrome (PJS) is a rare hereditary disease characterized by melanocytic macules of the lips, gastrointestinal hamartomatous polyps and an increased risk for many classes of cancer. The serine/threonine kinase LKB1 (also designated STK11) has been identified as the gene mutated in PJS. LKB1 activity increases upon the binding of a regulatory complex consisting of the STE20-related adaptor-alpha (STRAD alpha) pseudo kinase and the calcium binding protein 39 (MO25 alpha). STRAD and MO25 determine the subcellular localization of LKB1 by initiating its translocation from the nucleus to the cytoplasm, thus regulating the tumor suppressor activity of LKB1. The LKB1/STRAD/MO25 complex acts as an AMP-activated protein kinase kinase (AMPKK).Specifications
| CAB39 | |
| 0.3 mg/mL | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q06138, Q9Y376 | |
| 12283, 301574, 51719 | |
| A synthesized peptide derived from human CAB39/MO25 (aa 323-341). | |
| Antibody |
| 24GB6950 | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human, Mouse, Rat | |
| CAB39 | |
| AA408805; AA960512; C78372; Cab39; calcium binding protein 39; calcium binding protein, 39 kDa; calcium-binding protein 39; CGI-66; MO25; MO25alpha; Protein Mo25 | |
| CAB39 | |
| Primary | |
| Affinity chromatography |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title