missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CAB39 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA550709
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total CAB39 protein.
Peutz-Jeghers Syndrome (PJS) is a rare hereditary disease characterized by melanocytic macules of the lips, gastrointestinal hamartomatous polyps and an increased risk for many classes of cancer. The serine/threonine kinase LKB1 (also designated STK11) has been identified as the gene mutated in PJS. LKB1 activity increases upon the binding of a regulatory complex consisting of the STE20-related adaptor-alpha (STRAD alpha) pseudo kinase and the calcium binding protein 39 (MO25 alpha). STRAD and MO25 determine the subcellular localization of LKB1 by initiating its translocation from the nucleus to the cytoplasm, thus regulating the tumor suppressor activity of LKB1. The LKB1/STRAD/MO25 complex acts as an AMP-activated protein kinase kinase (AMPKK).
Specifications
| CAB39 | |
| Polyclonal | |
| Unconjugated | |
| CAB39 | |
| AA408805; AA960512; C78372; Cab39; calcium binding protein 39; calcium binding protein, 39 kDa; calcium-binding protein 39; CGI-66; MO25; MO25alpha; Protein Mo25 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 51719 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 2.9 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q9Y376 | |
| CAB39 | |
| Synthetic peptide corresponding to a region derived from internal residues of human calcium binding protein 39. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction