missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CA9 Human ELISA Kit
Sandwich ELISA Kit
Brand: Invitrogen™ EHCA9X10
Additional Details : Weight : 2.50000kg
Description
Carbonic anhydrase (CA) is an enzyme that assists rapid interconversion of carbon dioxide and water into carbonic acid, protons, and bicarbonate ions. It is abundant in all mammalian tissues. There are many genes that are inducible by hypoxia, via HIF-1 alpha. CA IX is one of the most inducible genes because of its stability and location within the membrane. Carbonic anhydrases have a widespread role in regulating pH in normal tissues, by regulating hydrogen ion (H+) flux. The pH is important in cell death under hypoxia, thus a blockade of CA IX results in increased cell death under hypoxia. Therefore, CA IX has become a reliable histochemical marker of hypoxia.Specifications
Q16790 | |
0.2 pg/mL | |
ELISA | |
Colorimetric Microplate Reader | |
CA9 | |
<10% | |
HRP | |
RUO | |
2°C to 8°C | |
1 hr 20 min |
0.205 to 50 pg/mL | |
Biotin | |
Plasma, Serum, Supernatant | |
768 | |
<12% | |
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
10 x 96 Tests | |
Plasma, 20 μL|Serum, 20 μL|Supernatant, 100 μL | |
Carbonic Anhydrase IX | |
4 hr 45 min |