missing translation for 'onlineSavingsMsg'
Learn More

Ca2+ channel agonist 1, MedChemExpress

Product Code. 16478048
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16478048

Brand: Medchem Express HY41076

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Ca2+ channel agonist 1 is an agonist of N-type Ca2+ channel and an inhibitor of Cdk2, with EC50s of 14.23 μM and 3.34 μM, respectively, and is used as a potential treatment for motor nerve terminal dysfunction.

Ca2+ channel agonist 1 is an agonist of N-type Ca2+ channel and an inhibitor of Cdk2, with EC50s of 14.23 μM and 3.34 μM, respectively, and is used as a potential treatment for motor nerve terminal dysfunction.

Specifications

Chemical Name or Material Ca2+ channel agonist 1
Physical Form Solid
CAS 1402821-24-2
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C19H26N6O
For Use With (Application) Neuroscience-Neuromodulation
Solubility Information DMSO : 50 mg/mL (141.06 mM; Need ultrasonic)
SMILES CC[C@@H](NC1=NC(NCC2=CC=CC=C2)=C3C(N(CCC)C=N3)=N1)CO
Molecular Weight (g/mol) 354.45
Formula Weight 354.45
Percent Purity 99.65%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.