missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CA19-9/Sialyl Lewis A Antibody (CA19.9/1390R), Janelia Fluor™ 646, Novus Biologicals™
Shop All Bio Techne ProductsDescription
CA19-9/Sialyl Lewis A Monoclonal antibody specifically detects CA19-9/Sialyl Lewis A in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | CA19-9/Sialyl Lewis A |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | CA19.9/1390R |
| Conjugate | Janelia Fluor 646 |
| Formulation | 50 mM Sodium Borate |
| Host Species | Rabbit |
| Immunogen | Purified human CA19-9/Sialyl Lewis A protein |
| Purification Method | Protein A purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?