missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CA150 Polyclonal Antibody
Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA564187
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Immunogen sequence: KIMQAKEDFK KMMEEAKFNP RATFSEFAAK HAKDSRFKAI EKMKDREALF NEFVAAARKK EKEDSKTRGE KIKSDFFELL SNHHLDSQSR WSKV Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
This gene encodes a nuclear protein that regulates transcriptional elongation and pre-mRNA splicing. The encoded protein interacts with the hyperphosphorylated C-terminal domain of RNA polymerase II via multiple FF domains, and with the pre-mRNA splicing factor SF1 via a WW domain. Alternative splicing results in multiple transcripts variants encoding different isoforms.
Especificaciones
| CA150 | |
| Polyclonal | |
| Unconjugated | |
| TCERG1 | |
| 2410022J09Rik; 2900090C16Rik; AI428505; CA150; CA150b; coactivator of 150 kD; co-activator of 150 kDa; FBP 28; FBP23; FBP28; Formin-binding protein 28; LOW QUALITY PROTEIN: transcription elongation regulator 1; MGC133200; p144; Taf2s; TATA box binding protein (TBP)-associated factor, RNA polymerase II, S, 150kD; TATA box-binding protein-associated factor 2S; TCERG1; Transcription elongation regulator 1; transcription elongation regulator 1 (CA150); Transcription elongation regulator 1-like protein; transcription factor CA150; transcription factor CA150b; Urn1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10915 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O14776 | |
| TCERG1 | |
| Recombinant protein corresponding to Human CA150. Recombinant protein control fragment (Product #RP-103574). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido