missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CA125/MUC16 Rabbit anti-Human, Clone: OCA125/2349R, Novus Biologicals™
Rabbit Monoclonal Antibody
£440.00
Specifications
| Antigen | CA125/MUC16 |
|---|---|
| Clone | OCA125/2349R |
| Concentration | 0.2 mg/mL |
| Dilution | Immunohistochemistry-Paraffin 1 to 2 μg/mL |
| Applications | Immunohistochemistry (Paraffin) |
Description
CA125/MUC16 Monoclonal specifically detects CA125/MUC16 in Human samples. It is validated for Immunohistochemistry-Paraffin.Specifications
| CA125/MUC16 | |
| 0.2 mg/mL | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cancer, Cellular Markers, Extracellular Matrix, Tumor Biomarkers | |
| 10 mM PBS with 0.05% BSA | |
| 94025 | |
| IgG | |
| Protein A or G purified |
| OCA125/2349R | |
| Immunohistochemistry-Paraffin 1 to 2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CA-125, CA125 ovarian cancer antigen, CA125MUC-16, FLJ14303, mucin 16, cell surface associated, mucin-16, Ovarian cancer-related tumor marker CA125, Ovarian carcinoma antigen CA125 | |
| Purified human CA125/MUC16 protein | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title