missing translation for 'onlineSavingsMsg'
Learn More

CA125/MUC16 Antibody (OCA125/2349R) - Azide and BSA Free, Novus Biologicals™

Product Code. 18360642 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18360642 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18360642 Supplier Novus Biologicals Supplier No. NBP308371

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

CA125/MUC16 Monoclonal specifically detects CA125/MUC16 in Human samples. It is validated for Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CA125/MUC16
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone OCA125/2349R
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry-Paraffin : 1-2 ug/mL
Formulation 10mM PBS
Gene Alias CA-125, CA125 ovarian cancer antigen, CA125MUC-16, FLJ14303, mucin 16, cell surface associated, mucin-16, Ovarian cancer-related tumor marker CA125, Ovarian carcinoma antigen CA125
Host Species Rabbit
Immunogen Purified human CA125/MUC16 protein
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, Cellular Markers, Extracellular Matrix, Tumor Biomarkers
Primary or Secondary Primary
Gene ID (Entrez) 94025
Target Species Human
Content And Storage Store at -20 to -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.