missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CA125/MUC16 Antibody (10F10), Novus Biologicals™
Rabbit Monoclonal Antibody
£260.00 - £406.00
Specifications
| Antigen | CA125/MUC16 |
|---|---|
| Clone | 10F10 |
| Dilution | ELISA, Immunohistochemistry 1:50-1:200 |
| Applications | ELISA, Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30030478
|
Novus Biologicals
NBP3-26127-50ul |
50 μL |
£260.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30037660
|
Novus Biologicals
NBP3-26127-100ul |
100 μL |
£406.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CA125/MUC16 Monoclonal antibody specifically detects CA125/MUC16 in Human samples. It is validated for ELISA, ImmunohistochemistrySpecifications
| CA125/MUC16 | |
| ELISA, Immunohistochemistry 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CA-125, CA125 ovarian cancer antigen, CA125MUC-16, FLJ14303, mucin 16, cell surface associated, mucin-16, Ovarian cancer-related tumor marker CA125, Ovarian carcinoma antigen CA125 | |
| A synthesized peptide derived from Human CA125/MUC16 [UniProt Q8WXI7] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 10F10 | |
| ELISA, Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Cancer, Cellular Markers, Extracellular Matrix, Tumor Biomarkers | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 94025 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title