missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CA125 Monoclonal Antibody (X325)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA124603
This item is not returnable.
View return policy
Description
MA1-24603 detects MUC16 from human samples.
CA 125 determinant is present on a high molecular weight, mucin like glycoprotein of high molecular weight. CA 125 has been found on frozen sections of amnion and derivatives of coelomic and mullerian epithelium, including pleura, pericardium and peritoneum. In adult tissues, epithelial cells of fallopian tube, endometrium and endocervix; pancreas, colon, gall bladder, stomach, kidney, apocrine sweat gland and mammary gland. It is also found in mesothelial cell lining of pleura, pericardium and peritoneum. It is found in ovarian tumors of serous, endometrioid or clear cell types and adenocarcinomas of mullerian type.
Specifications
| CA125 | |
| Monoclonal | |
| 5.3 mg/mL | |
| 50mM sodium citrate with 150mM NaCl and 0.09% sodium azide; pH 7.5 | |
| Q8WXI7 | |
| MUC16 | |
| CA125 antigen purified from human ovarian carcinoma. | |
| 500 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| X325 | |
| Unconjugated | |
| MUC16 | |
| 1110008I14Rik; Ca125; CA-125; CA125 ovarian cancer antigen; ca-ovarian; FLJ14303; Gm1480; MUC16; MUC-16; mucin 16; mucin 16, cell surface associated; mucin-16; Ovarian cancer-related tumor marker CA125; ovarian carcinoma antigen CA125; RGD1308422 | |
| Mouse | |
| Protein A | |
| RUO | |
| 94025 | |
| 4°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction