missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CA11 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15435-1-AP-150UL
This item is not returnable.
View return policy
Description
Does not have a catalytic activity.Specifications
| CA11 | |
| Polyclonal | |
| Unconjugated | |
| CA11 | |
| CA RP II, CA RP XI, CA XI, CA11, carbonic anhydrase XI, CARP 2, CARP XI, CARP2, CARPX1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 12348, 308588, 770 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.25 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O70354, O75493 | |
| CA11, Car11 | |
| CA11 Fusion Protein Ag7712 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction