missing translation for 'onlineSavingsMsg'
Learn More

Ca, Mg, Mo, P, Zn @ 2000μg/g in 75 cSt Oil, VHG™

Product Code. 17990726
Change view
Click to view available options
Quantity:
200 g
400 g
Unit Size:
200g
400g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17990726 400 g 400g
17973956 200 g 200g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17990726 Supplier VHG Labs Supplier No. VHGMA4+MO2000400G
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

Ca, Mg, Mo, P, Zn @ 2000μg/g in 75 cSt Oil
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Calcium/Magnesium/Molybdenum/Phosphorus/Zinc
CAS 7440-70-2, 7439-95-4, 7439-98-7, 7723-14-0, 7440-66-6
Methods ICP
Analyte or Component Names Calcium/Magnesium/Molybdenum/Phosphorus/Zinc
Shipping Condition Room Temperature
Certifications/Compliance ISO 17034
Solvent or Matrix 75 cSt Oil
Quantity 400 g
Concentration or Composition (by Analyte or Components) Calcium 2000 μg/g/Magnesium 2000 μg/g/Molybdenum 2000 μg/g/Phosphorus 2000 μg/g/Zinc 2000 μg/g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.