missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ca, Mg, Mo, P, Zn @ 2000μg/g in 75 cSt Oil, VHG™
Click to view available options
Quantity:
200 g
400 g
Unit Size:
200g
400g
Description
Ca, Mg, Mo, P, Zn @ 2000μg/g in 75 cSt Oil
Specifications
Specifications
| Chemical Name or Material | Calcium/Magnesium/Molybdenum/Phosphorus/Zinc |
| CAS | 7440-70-2, 7439-95-4, 7439-98-7, 7723-14-0, 7440-66-6 |
| Methods | ICP |
| Analyte or Component Names | Calcium/Magnesium/Molybdenum/Phosphorus/Zinc |
| Shipping Condition | Room Temperature |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | 75 cSt Oil |
| Quantity | 200 g |
| Concentration or Composition (by Analyte or Components) | Calcium 2000 μg/g/Magnesium 2000 μg/g/Molybdenum 2000 μg/g/Phosphorus 2000 μg/g/Zinc 2000 μg/g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction