missing translation for 'onlineSavingsMsg'
Learn More

CA-170, MedChemExpress

Product Code. 16420729
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16420729

Brand: Medchem Express HY101093

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress CA-170 is an orally delivered dual inhibitor of VISTA and PD-L1. CA-170 exhibits potent rescue of proliferation and effector functions of T cells inhibited by PD-L1/L2 and VISTA with selectivity over other immune checkpoint proteins as well as a broad panel of receptors and enzymes.

CA-170 is an orally delivered dual inhibitor of VISTA and PD-L1. CA-170 exhibits potent rescue of proliferation and effector functions of T cells inhibited by PD-L1/L2 and VISTA with selectivity over other immune checkpoint proteins as well as a broad panel of receptors and enzymes.

Specifications

Chemical Name or Material CA-170
Physical Form Solid
CAS 1673534-76-3
Color Light Yellow
Quantity 1 mg
Purity Grade Notes Research
Molecular Formula C12H20N6O7
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : 28.5 mg/mL (79.10 mM; Need ultrasonic and warming)
SMILES C[C@@H](O)[C@@H](C(O)=O)NC(N[C@H](C1=NC([C@@H](N)CO)=NO1)CC(N)=O)=O
Molecular Weight (g/mol) 360.32
Formula Weight 360.32
Percent Purity 95.14%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.