missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ CA 125 (Ovarian Cancer Antigen) Ab-1, Mouse Monoclonal Antibody

Obtain accurate and reproducible results in immunohistochemistry experiments with Epredia™ CA 125 (Ovarian Cancer Antigen) Ab-1, Mouse Monoclonal Antibody.

£97.05 - £345.00

Specifications

Antigen CA 125 (Ovarian Cancer Antigen) Ab-1
Clone Ov185:1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 5
Product Code Brand Quantity Form Price Quantity & Availability  
Product Code Brand Quantity Form Price Quantity & Availability  
12619126
View Documents
Epredia™
MS-1151-S0
100 μL Concentrated This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12629126
View Documents
Epredia™
MS-1151-S1
500 μL Concentrated
ONLINE EXCLUSIVE PRICE
£278.00
£260.73 / 500µL
Save £17.27
6% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
12609126
View Documents
Epredia™
MS-1151-S
1 mL Concentrated This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12689116
View Documents
Epredia™
MS-1151-R7
7 mL Prediluted
ONLINE EXCLUSIVE PRICE
£229.00
£214.20 / 7mL
Save £14.80
6% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
12699116
View Documents
Epredia™
MS-1151-RQ
12 mL Ready-to-Use This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

As CA125 (Ovarian Cancer Antigen) is a tumour marker usually associated with ovarian epithelial malignancies. Serum assays are widely used to detect this protein in the monitoring of ovarian cancers. However, CA125 antigen may also be detected by immunohistochemistry and expression has been found in neoplasms such as seminal vesicle carcinoma and anaplastic lymphoma. CA125 antigen is not exclusively found in malignant tumours.

Host Species: Mouse

Clone: Ov185:1

Isotype: IgG1

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Partially purified mucin fraction from a pool of cancer tissue from patients with epithelial ovarian cancer

Positive Control: Ovarian CA

Cellular Localization: Extracellular membrane-associated

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Specifications

CA 125 (Ovarian Cancer Antigen) Ab-1
Immunohistochemistry (Paraffin)
Unconjugated
IVD
Partially purified mucin fraction from a pool of cancer tissue from patients with epithelial ovarian cancer
Ov185:1
Monoclonal
Mouse
Oncogenesis
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.