Learn More
Epredia™ Lab Vision™ CA 125 Ab-2, Mouse Monoclonal Antibody
Obtain accurate, reproducible results in immunohistochemistry procedures with Epredia™ CA 125 Ab-2, Mouse Monoclonal Antibody.
£530.40 - £708.90
Specifications
| Antigen | CA 125 Ab-2 |
|---|---|
| Clone | M11 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
Description
CA 125 determinant is present on a high molecular weight, mucin like glycoprotein of high molecular weight. CA 125 has been found on frozen sections of amnion and derivatives of coelomic and mullerian epithelium, including pleura, pericardium and peritoneum. In adult tissues, epithelial cells of fallopian tube, endometrium and endocervix; pancreas, colon, gall bladder, stomach, kidney, apocrine sweat gland and mammary gland. It is also found in mesothelial cell lining of pleura, pericardium and peritoneum. It is found in ovarian tumors of serous, endometrioid or clear cell types and adenocarcinomas of mullerian type.Host Species: Mouse
Clone: M11
Isotype: IgG1/κ
Speices Reactivity: Human. Others not known.
Epitope: Different than clone OC125
Immunogen: CA 125 antigen derived from ascites fluid
Molecular Weight: >200kDa
Positive Control: Breast, ovarian, endometrial, colon and lung CA
Cellular Localization: Extracellular membrane-associated
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Note:
USA: RUO; Int'l: RUO
Specifications
| CA 125 Ab-2 | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| IgG2a κ |
| M11 | |
| Monoclonal | |
| Mouse | |
| Oncogenesis | |
| CA 125 antigen derived from ascites fluid | |
| Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.