missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C9orf89 Polyclonal Antibody

C9orf89 Polyclonal Antibody
Brand: Invitrogen PA589068
This item is not returnable.
View return policy
Description
C9orf89 Polyclonal Antibody for Western Blot
Specifications
| C9orf89 | |
| Polyclonal | |
| Unconjugated | |
| CARD19 | |
| 1110007C09Rik; AI851695; Bcl10-interacting CARD protein; Bcl10-interacting protein with CARD; BinCARD; C9orf89; CARD19; caspase recruitment domain family member 19; caspase recruitment domain family, member 19; caspase recruitment domain-containing protein 19; RIKEN cDNA 1110007C09 gene | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 68480, 84270 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.48 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q96LW7, Q9D1I2 | |
| CARD19 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-100 of human CARD19 (NP_1156863). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction