missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ C9orf72 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5114512
This item is not returnable.
View return policy
Description
Positive controls that may be used include: A20 Cell Lysate, A-20 Cell Slide.
C9orf72 may play a role in endosomal trafficking and autophagy. It is known to interact with HNRNPA1, HNRNPA2B1, UBQLN2, and small Rab GTPSases. C9orf72 localizes in the nucleus, cytoplasm, endosome, lysosome, and autophagosome. Mutations in this gene can result in frontotemporal dementia and/or amyotrophic lateral sclerosis.
Specifications
| C9orf72 | |
| Polyclonal | |
| Unconjugated | |
| C9orf72 | |
| 3110043O21Rik; AI840585; ALSFTD; C9orf72; chromosome 9 open reading frame 72; FTDALS; FTDALS1; Guanine nucleotide exchange C9orf72; Guanine nucleotide exchange C9orf72 homolog; protein C9orf72; protein C9orf72 homolog; RGD1359108; RIKEN cDNA 3110043O21 gene; similar to RIKEN cDNA 3110043O21 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 203228, 313155, 73205 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| Q66HC3, Q96LT7 | |
| C9orf72, RGD1359108 | |
| ALSFTD antibody was raised against a 19 amino acid peptide from near the carboxy terminus of human ALSFTD. The immunogen is located within amino acids 400 - 450 of ALSFTD. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction