missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C9orf142 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27134-1-AP-20UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene plays a role in the nonhomologous end joining (NHEJ) pathway of DNA double-strand break repair. The encoded protein may function to stabilize the Ku70/Ku80 heterodimer to facilitate the assembly and maintain the stability of the NHEJ complex.Specifications
| C9orf142 | |
| Polyclonal | |
| Unconjugated | |
| C9orf142 | |
| C9orf142 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 286257 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.47 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9BUH6 | |
| C9orf142 | |
| C9orf142 Fusion Protein Ag24910 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction