missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ C9orf117 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA554420
This item is not returnable.
View return policy
Description
Immunogen sequence: EEVTDKFTLL EEQVRKQENE FRDYAYNLEK KSVLDKDRLR KEIIQRVNLV ANEFHKVTTN RMWETTKRAI KENNGITLQM ARVSQQGMKL LQENEQL Highest antigen sequence identity to the following orthologs: Mouse - 70%, Rat - 74%.
Specifically required during spermatogenesis for flagellum morphogenesis and sperm motility. May be required to suppress the formation of supernumerary axonemes and ensure a correct ultrastructure.
Specifications
| C9orf117 | |
| Polyclonal | |
| Unconjugated | |
| CFAP157 | |
| 1700019L03Rik; C25H9orf117; C9H9orf117; C9orf117; Cfap157; cilia and flagella associated protein 157; cilia- and flagella-associated protein 157; LOC499781; LOW QUALITY PROTEIN: uncharacterized protein C9orf117 homolog; RIKEN cDNA 1700019L03 gene; similar to CG17122-PA; uncharacterized protein C9orf117; uncharacterized protein C9orf117 homolog | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 286207 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 1.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q5JU67 | |
| CFAP157 | |
| Recombinant protein corresponding to Human C9orf117. Recombinant protein control fragment (Product #RP-93025). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction