missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ C9orf116 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5111598
This item is not returnable.
View return policy
Description
Immunogen sequence: MAEECPRACA EPVAPKATAP PERTSDYYRV SADLPGRFNN PGWFRGYRTQ KAVSVYRTSN Highest antigen sequence indentity to the following orthologs: Rat - 70%, Mouse - 58%.
C9orf116 (Chromosome 9 Open Reading Frame 116) is a Protein Coding gene. An important paralog of this gene isC15orf65.
Specifications
| C9orf116 | |
| Polyclonal | |
| Unconjugated | |
| C9orf116 | |
| C9orf116; chromosome 9 open reading frame 116; p53-induced expression 1 in Rb-/- cells; p53-induced expression in RB-null cells protein 1; pierce 1; PIERCE1; RbEST47; UPF0691 protein C9orf116 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 138162 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q5BN46 | |
| C9orf116 | |
| Recombinant Protein corresonding to Human C9orf116. Recombinant protein control fragment (Product #RP-108507). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction