Learn More
Invitrogen™ C8orf34 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA5112951
Product Code. 17120814
  • £400.00 / 100µg
Description
C8orf34 Polyclonal Antibody for Western Blot, IHC (P), ELISA
This gene encodes a protein that is related to the cyclic AMP dependent protein kinase regulators. Naturally occurring mutations in this gene are associated with an increased risk for severe toxicities, such as diarrhea and neutropenia, in patients undergoing chemotherapeutic treatment.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
C8orf34
Polyclonal
Unconjugated
C8orf34
C8orf34; chromosome 8 open reading frame 34; protein VEST-1; Uncharacterized protein C8orf34; VEST1; VEST-1; vestibule 1; vestibule-1 protein
Rabbit
Protein G
RUO
116328
-20°C or -80°C if preferred
Liquid
ELISA, Immunohistochemistry (Paraffin), Western Blot
2.5 mg/mL
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Q49A92
C8orf34
Recombinant Human Uncharacterized protein C8orf34 protein (1-300AA).
100 μg
Primary
Human, Mouse