missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ C8G Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody
£161.00 - £333.00
Specifications
| Antigen | C8G |
|---|---|
| Concentration | 0.5 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17930223
![]() |
Invitrogen™
PA5145303 |
200 μL |
£333.00
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17919302
|
Invitrogen™
PA5145304 |
20 μL |
£161.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
C8G is one of the three polypeptides that constitute C8, a component of the complement system. C8 participates in the formation of Membrane Attack Complex (MAC). Patients with deficiency in C8 are vulnerable to certain bacteria infection.Specifications
| C8G | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Pig | |
| P07360 | |
| 100037955, 733 | |
| Recombinant Human C8g protein, Lys39-Arg202 (Accession #P07360). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
| 0.5 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
| C8C; C8G; complement component 8, gamma polypeptide; complement component C8 gamma chain | |
| C8G | |
| IgG | |
| Antigen affinity chromatography, Protein A | |
| C8G |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title