missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ C8B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA597523
This item is not returnable.
View return policy
Description
C8B Polyclonal Antibody for Western Blot, IHC (P), ELISA
C8 beta is one of the three subunits that comprise the component 8 (C8) of the complement system. C8 participates in the formation of Membrane Attack Complex that results in the lysis of cells. Patients with C8B deficiency are prone to bacteria infection.
Specifications
| C8B | |
| Polyclonal | |
| Unconjugated | |
| C8B | |
| 4930439B20Rik; AI595927; C82; C8b; complement C8 beta chain; complement component 8 subunit beta; complement component 8, beta polypeptide; complement component 8, beta subunit; complement component C8 beta chain; complement component C8 beta subunit; MGC137114 protein; MGC163447 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 110382, 732 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 0.31 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P07358, Q8BH35 | |
| C8B | |
| Recombinant Human Complement component C8 beta chain protein (55-350AA). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction