missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C8B Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92538-0.1ml
This item is not returnable.
View return policy
Description
C8B Polyclonal antibody specifically detects C8B in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| C8B | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| complement component 8 subunit beta, complement component 8, beta polypeptide, MGC163447 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 55-350 of human C8B (NP_000057.2). SVDVTLMPIDCELSSWSSWTTCDPCQKKRYRYAYLLQPSQFHGEPCNFSDKEVEDCVTNRPCRSQVRCEGFVCAQTGRCVNRRLLCNGDNDCGDQSDEANCRRIYKKCQHEMDQYWGIGSLASGINLFTNSFEGPVLDHRYYAGGCSPHYILNTRFRKPYNVESYTPQTQGKYEFILKEYESYSDFERNVTEKMASKSGFSFGFKIPGIFELGISSQSDRGKHYIRRTKRFSHTKSVFLHARSDLEVAHYKLKPRSLMLHYEFLQRVKRLPLEYSYGEYRDLFRDFGTHYITEAVL | |
| 0.1 mL | |
| Cardiovascular Biology, Immunology, Innate Immunity, Neurodegeneration, Neuroscience | |
| 732 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction