missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ C8A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA568482
This item is not returnable.
View return policy
Description
Predicted to react with Human samples.
C8A is a component of the complement system and contains three polypeptide subunits: alpha, beta and gamma. C8A, in particular, contains the alpha subunit of C8 and is a constituent of the membrane attack complex (MAC) that plays a key role in the innate and adaptive immune response by forming pores in the plasma membrane of target cells. C8A inserts into the target membrane, but does not form pores by itself. Disease associated with C8A include Type 1 C8 deficiency and immunodeficiency due to C8A deficiency.
Specifications
| C8A | |
| Polyclonal | |
| Unconjugated | |
| C8A | |
| C8a; complement component 8 alpha subunit; complement component 8 subunit alpha; complement component 8, alpha polypeptide; complement component C8 alpha chain | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 731 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P07357 | |
| C8A | |
| Synthesized peptide derived from human C8A [aa 201-250]. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction