missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ C7orf57 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA554372
This item is not returnable.
View return policy
Description
Immunogen sequence: ISNGYKDEWL QQQQRADSDK RTPKTSRASV LSQSPRDLEG PQDAARLQDA EASEGPEDTP GPEESVSAST PA Highest antigen sequence identity to the following orthologs: Mouse - 59%, Rat - 57%.
C7orf57 is a protein coding gene.
Specifications
| C7orf57 | |
| Polyclonal | |
| Unconjugated | |
| C7orf57 | |
| C7orf57; chromosome 7 open reading frame 57; uncharacterized protein C7orf57 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 136288 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8NEG2 | |
| C7orf57 | |
| Recombinant protein corresponding to Human C7orf57. Recombinant protein control fragment (Product #RP-92794). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction