missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ C7orf50 Monoclonal Antibody (K16257_13H2), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA555965
This item is not returnable.
View return policy
Description
Sequence of this protein is as follows: VTEKKNKKLK KASAEGPLLG PEAAPSGEGA GSKGEAVLRP GLDAEPELSP EEQRVLERKL KKERKKEERQ RLREAGLVAQ HPPARRSGAE LALDYLCRWA QKHKNWRFQK TRQTWLLLHM YDSDKVPDEH FSTLLAYLEG LQGRARELTV QKAEALMREL DEEGSDPPLP
C7orf50 gene ontology annotations related to this gene include RNA binding.
Specifications
| C7orf50 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| Q9BRJ6 | |
| C7orf50 | |
| Recombinant human C7orf50 (immunogen range is 11-180). | |
| 50 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| K16257_13H2 | |
| Unconjugated | |
| C7orf50 | |
| C7orf50; chromosome 7 open reading frame 50; FP15621; MGC11257; Uncharacterized protein C7orf50; YCR016W | |
| Mouse | |
| Protein A | |
| RUO | |
| 84310 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction