missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ C7orf29 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA565976
This item is not returnable.
View return policy
Description
Immunogen sequence: MVVREASAAQA SLSQVLPQLR YLHIFLEQVH THFQEQSVGE RGAAIQLAEG LARQLCTDCQ LNKLFYREEF VLATLLDP Highest antigen sequence identity to the following orthologs - mouse 32%, rat 32%.
ZBED6CL is a protein coding gene.
Specifications
| C7orf29 | |
| Polyclonal | |
| Unconjugated | |
| ZBED6CL | |
| C7orf29; ZBED6 C-terminal like; ZBED6 C-terminal-like protein; ZBED6CL | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 113763 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96FA7 | |
| ZBED6CL | |
| Recombinant Human C7orf29. Recombinant protein control fragment (Product #RP-106665). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction