missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C7orf10 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21589-1-AP-150UL
This item is not returnable.
View return policy
Description
Russel-Silver syndrome candidate,dermal papilla derived protein 13,hypothetical protein LOC79783Specifications
| C7orf10 | |
| Polyclonal | |
| Unconjugated | |
| SUGCT | |
| C7orf10, DERP13, ORF19 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 192136, 361253, 79783 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.17 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q68FU4, Q7TNE1, Q9HAC7 | |
| Sugct | |
| C7orf10 Fusion Protein Ag16159 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction