Learn More
Invitrogen™ C7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5120912
Description
Positive test controls include: Mouse plasma, Rat plasma. The target is usually found in the following locations: Secreted. Immunogen sequence: ASSPVNCQWD FYAPWSECNG CTKTQTRRRS VAVYGQYGGQ PCVGNAFETQ SCEPTRGCPT EEGCGERFRC FSGQCISKSL VCNGDSDCDE DSADEDRCED SERRPSCDID KPPPNIELTG NGYNELTGQF RNRVINTKSF GGQCRKVFSG DGKDFYRLSG NVLSYTFQVK INNDFNYEFY NSTWSYVKHT STEHTSSSRK RSFFRSSSSS SRSYTSHTNE IHKGKSYQLL VVENTVEVAQ FINNNPEFL.
C7 is a component of the complement system. It participates in the formation of Membrane Attack Complex (MAC). People with C7 deficiency are prone to bacterial infection.
Specifications
| C7 | |
| Polyclonal | |
| Unconjugated | |
| C7 | |
| A530026G17; C7; complement component 7; complement component C7; Gm1238 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 109828, 117517, 730 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 0.79 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| P10643 | |
| C7 | |
| Synthetic peptide. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.