missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ C6orf165 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA567233

Product Code. 15939904

  • £476.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: GVLSNLFTHIQ PFLGAHELYF PERVMQCHLN GATVKTDVCR MKEHMEDRVN VADFRKLEWL FPETTANFDK LLIQYRGFCA YTFAATDGLL L Highest antigen sequence identity to the following orthologs - mouse 70%, rat 70%.

CFAP206 may regulate cilium motility through its role in the assembly of the axonemal radial spokes.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

C6orf165
Polyclonal
Unconjugated
CFAP206
1700003M02Rik; AU024269; C6orf165; Cfap206; cilia and flagella associated protein 206; Cilia- and flagella-associated protein 206; dJ382I10.1; RGD1310641; RIKEN cDNA 1700003M02 gene; similar to hypothetical protein; UPF0704 protein C6orf165; UPF0704 protein C6orf165 homolog
Rabbit
Antigen Affinity Chromatography
RUO
154313
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry
0.2 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q8IYR0
CFAP206
Recombinant Human C6orf165. Recombinant protein control fragment (Product #RP-107869).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.